ContactUsLinkComponent
A gastroprotective histamine H2 receptor antagonist (pKi = 7.1) that decreases gastric acid secretion and inhibits intracellular cAM P production (IC50 = 1 µM) 1 * 5 g
Catalog # CAYM16939-5
Supplier:  Cayman Chemical
A gastroprotective histamine H2 receptor antagonist (pKi = 7.1) that decreases gastric acid secretion and inhibits intracellular cAM P production (IC50 = 1 µM) 1 * 5 g
Catalog # CAYM16939-5
Supplier:  Cayman Chemical