ContactUsLinkComponent
A steroidal alkaloid that suppresses NF-B signaling in LPS-stimulated macrophages, blocking induced expression of iNOS and COX-2 1 * 10 mg
Catalog # CAYM16938-10
Supplier:  Cayman Chemical
A steroidal alkaloid that suppresses NF-B signaling in LPS-stimulated macrophages, blocking induced expression of iNOS and COX-2 1 * 10 mg
Catalog # CAYM16938-10
Supplier:  Cayman Chemical