ContactUsLinkComponent
A broad-spectrum, indole-based antiviral compound that blocks viral fusion with target membranes, prohibiting viral entry into cells 1 * 100 mg
Catalog # CAYM16933-100
Supplier:  Cayman Chemical
A broad-spectrum, indole-based antiviral compound that blocks viral fusion with target membranes, prohibiting viral entry into cells 1 * 100 mg
Catalog # CAYM16933-100
Supplier:  Cayman Chemical