ContactUsLinkComponent
An ATP-competitive, pan-Akt kinase inhibitor (IC50s = 2, 13, and 9 nM for Akt1, 2, and 3, respectively) that inhibits proliferation and induces apoptosis in various human tumor cells in vitro and in xenografts in immunocompromised mice 1 * 10 mg
Catalog # CAYM16891-10
Supplier:  Cayman Chemical
An ATP-competitive, pan-Akt kinase inhibitor (IC50s = 2, 13, and 9 nM for Akt1, 2, and 3, respectively) that inhibits proliferation and induces apoptosis in various human tumor cells in vitro and in xenografts in immunocompromised mice 1 * 10 mg
Catalog # CAYM16891-10
Supplier:  Cayman Chemical