ContactUsLinkComponent
An agonist of GPR81 (EC50 = 16 µM) that is inactive against the related GPR109a (HCAR2) receptor 1 * 500 mg
Catalog # CAYM16795-500
Supplier:  Cayman Chemical
An agonist of GPR81 (EC50 = 16 µM) that is inactive against the related GPR109a (HCAR2) receptor 1 * 500 mg
Catalog # CAYM16795-500
Supplier:  Cayman Chemical