ContactUsLinkComponent
A potent, cell-permeable inhibitor of SET7/9 (IC50 = 2 nM) that demonstrates greater than 1,000-fold selectivity over a panel of 18 other methyltransferases 1 * 5 mg
Catalog # CAYM14678-5
Supplier:  Cayman Chemical
A potent, cell-permeable inhibitor of SET7/9 (IC50 = 2 nM) that demonstrates greater than 1,000-fold selectivity over a panel of 18 other methyltransferases 1 * 5 mg
Catalog # CAYM14678-5
Supplier:  Cayman Chemical
Frequently Bought Together