ContactUsLinkComponent
A blocker of KCNQ channels that potently inhibits KCNQ1 and 2 homomeric channels (IC50 = 0.75 and 0.71 µM, respectively) as well as KCNQ2+3 heteromultimers (IC50 = 0.6 µM) 1 * 5 mg
  CAYM14582-5
A blocker of KCNQ channels that potently inhibits KCNQ1 and 2 homomeric channels (IC50 = 0.75 and 0.71 µM, respectively) as well as KCNQ2+3 heteromultimers (IC50 = 0.6 µM) 1 * 5 mg
  CAYM14582-5