ContactUsLinkComponent
5 663 010 Vacuum Safety bottle, WITOFORM-plasic-coated, 10000 ml, with 3 necks, CN GL 25, SN GL18, with suction tube, safety valve, pressure gauge, air-suction tube, PTFE needle valve stopcock 1 * 1 items
Catalog # WITG5.663.010
Supplier:  witeg Labortechnik
5 663 010 Vacuum Safety bottle, WITOFORM-plasic-coated, 10000 ml, with 3 necks, CN GL 25, SN GL18, with suction tube, safety valve, pressure gauge, air-suction tube, PTFE needle valve stopcock 1 * 1 items
Catalog # WITG5.663.010
Supplier:  witeg Labortechnik
Frequently Bought Together