EC HPLC column EC 150/3 NUCLEOSIL 100-3 Protect I length: 150 mm, ID: 3 mm pack of 1 1 * 1 items
MANA720541.30
EC HPLC column EC 150/3 NUCLEOSIL 100-3 Protect I length: 150 mm, ID: 3 mm pack of 1 1 * 1 items
MANA720541.30
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection