ContactUsLinkComponent
Rabbit anti-T7 is a primary antibody which binds to T7 and is directly conjugated to DyLight® 650. This is of advantage to shorten a ssay time by no need to use a secondary antibody to T7. 1 * 100 µG
Catalog # AGRIAS152978
Supplier:  Agrisera
Rabbit anti-T7 is a primary antibody which binds to T7 and is directly conjugated to DyLight® 650. This is of advantage to shorten a ssay time by no need to use a secondary antibody to T7. 1 * 100 µG
Catalog # AGRIAS152978
Supplier:  Agrisera