ContactUsLinkComponent
Rabbit anti-S is a primary antibody which binds to S and is directly conjugated to DyLight® 650. This is of advantage to shorten ass ay time by no need to use a secondary antibody to S. 1 * 100 µG
Catalog # AGRIAS152970
Supplier:  Agrisera
Rabbit anti-S is a primary antibody which binds to S and is directly conjugated to DyLight® 650. This is of advantage to shorten ass ay time by no need to use a secondary antibody to S. 1 * 100 µG
Catalog # AGRIAS152970
Supplier:  Agrisera