ContactUsLinkComponent
Rabbit anti-V5 is a primary antibody which binds to V5 and is directly conjugated to DyLight® 488. This is of advantage to shorten a ssay time by no need to use a secondary antibody to V5. 1 * 100 µG
Catalog # AGRIAS152966
Supplier:  Agrisera
Rabbit anti-V5 is a primary antibody which binds to V5 and is directly conjugated to DyLight® 488. This is of advantage to shorten a ssay time by no need to use a secondary antibody to V5. 1 * 100 µG
Catalog # AGRIAS152966
Supplier:  Agrisera