Rabbit anti-V5 is a primary antibody which binds to V5 and is directly conjugated to ALP, Alkaline Phosphatase. This is of advantage to shorten assay time by no need to use a secondary antibody to V5. 1 * 100 µG
Catalog # AGRIAS152964
Supplier: Agrisera