ContactUsLinkComponent
Mouse anti-c-myc is a primary antibody which binds to c-myc tag and is directly conjugated to DyLight® 488. 1 * 100 µG
Catalog # AGRIAS152945
Supplier:  Agrisera
Mouse anti-c-myc is a primary antibody which binds to c-myc tag and is directly conjugated to DyLight® 488. 1 * 100 µG
Catalog # AGRIAS152945
Supplier:  Agrisera