Mouse anti-c-myc is a primary antibody which binds to c-myc and is directly conjugated to ALP, Alkaline Phosphatase. This is of adva ntage to shorten assay time by no need to use a secondary antibody to c-myc. 1 * 100 µG
Catalog # AGRIAS152940
Supplier: Agrisera