ContactUsLinkComponent
Fibroblast growth factor-21 (FGF21) belongs to the large FGF family which has at least 23 members. Along with FGF-19/15 and F GF-23, FGF-21 is categorized as a member of the atypical FGF subfamily, as it must be complexed to the Klotho co-rece 1 * 1 mg
Catalog # GENSZ03290-1
Supplier:  Genscript
Fibroblast growth factor-21 (FGF21) belongs to the large FGF family which has at least 23 members. Along with FGF-19/15 and F GF-23, FGF-21 is categorized as a member of the atypical FGF subfamily, as it must be complexed to the Klotho co-rece 1 * 1 mg
Catalog # GENSZ03290-1
Supplier:  Genscript