ContactUsLinkComponent
Safety storage container stainless steel. Capacity 50 L. Made of high-quality stainless steel 1.4301 / 1.4571 with thickness of sides of 1,5 mm. With screw cap and pressure-relief valve. Height 600 mm. Thread 2'' (TPI). Dia. 380 mm 1 * 1 items
Catalog # BURK2601-5050
Supplier:  BURKLE
Safety storage container stainless steel. Capacity 50 L. Made of high-quality stainless steel 1.4301 / 1.4571 with thickness of sides of 1,5 mm. With screw cap and pressure-relief valve. Height 600 mm. Thread 2'' (TPI). Dia. 380 mm 1 * 1 items
Catalog # BURK2601-5050
Supplier:  BURKLE
Frequently Bought Together