BIOCHEMICAL OXYGEN DEMAND (BOD) @ 200 MG/L IN H2O 1 * 100 mL
Catalog # PROOVHGBOD200A-100
Supplier: LGC Standards PROMOCHEM
BIOCHEMICAL OXYGEN DEMAND (BOD) @ 200 MG/L IN H2O 1 * 100 mL
Catalog # PROOVHGBOD200A-100
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
BUFFER PH4/7/10 3X500ML 1 * 3 items
Supplier: HAMILTON BONADUZ
UV/Visible standards, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals