BENZOPHENONE (DIPHENYLMETHANONE) 1 * 10 mg
Catalog # PROOLGCFOR0630.01
Supplier: LGC Standards PROMOCHEM
BENZOPHENONE (DIPHENYLMETHANONE) 1 * 10 mg
Catalog # PROOLGCFOR0630.01
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Sulfite in Water, Certified Reference Material, Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Hanna
SODIUM VALPROATE EP 1 * 25 g
Supplier: Spectrum Chemical
PH BUFFER KIT PH 4 ,7 , 10 1 * 1 KIT
Supplier: YSI ENVIRONMENTAL
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Sulphomolybdic reagent R2 Reag. Ph. Eur. 1086400
Supplier: VWR Chemicals
Phosphomolybdotungstic reagent dilute Reag. Ph. Eur. 1065001
Supplier: VWR Chemicals
Supelco® Organic Reference Standard, [6]-Shogaol
Supplier: Merck
Buffer Solutions
Supplier: Thermo Fisher Scientific