ContactUsLinkComponent
10µG/G AL, CR, CU, FE, PB, MG, NI, SI, AG, NA, SN, TI IN HYDROCARBON OIL 1 * 1 items
Catalog # PROOG-V12-10-200G
10µG/G AL, CR, CU, FE, PB, MG, NI, SI, AG, NA, SN, TI IN HYDROCARBON OIL 1 * 1 items
Catalog # PROOG-V12-10-200G
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.