DETERMINATION OF MERCAPTAN SULFUR FOR UOP 163 AND ASTM D3227 - S @ 30µG/G - 6 X 20 ML AMPOULES 1 * 1 items
Catalog # PROOGUOP16330-6X20
Supplier: LGC Standards PROMOCHEM
DETERMINATION OF MERCAPTAN SULFUR FOR UOP 163 AND ASTM D3227 - S @ 30µG/G - 6 X 20 ML AMPOULES 1 * 1 items
Catalog # PROOGUOP16330-6X20
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
Accessories for pH/°C meter, handheld
Supplier: Testo
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
Bisphenol A, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
C TECHNOLOGIES VPE MEASUREMENT STANDARD 1 * 1 KIT
Supplier: MARKET SOURCE PART PROCESS
VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Supplier: VWR Chemicals
Supelco® Organic Reference Standard, [6]-Shogaol
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion