GLYCOPYRRONIUM BROMIDE 1 * 10 mg
Catalog # EDQMY0001308
Supplier: EDQM
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
UV/Visible standards, Supelco®
Supplier: Merck
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Supplier: VWR Chemicals
Supelco® COD CRM, standard solutions
Supplier: Merck
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR®, Fluoride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Ammonia solution 17% Reag. Ph. Eur. 1004701
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals