ROCURONIUM BROMIDE 1 * 10 mg
Catalog # EDQMY0001580
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Ammonia solution 6 mol/l (6 N) Reag. Ph. Eur. 1004702
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals