ContactUsLinkComponent
Additional reinforced perforated stainless steel shelf, max. loading 60 kg with guide bars and fixing screws (only in connection wi th option K1) 1 * 1 items
Catalog # MEMMB32191UF750TS
Supplier:  MEMMERT
Additional reinforced perforated stainless steel shelf, max. loading 60 kg with guide bars and fixing screws (only in connection wi th option K1) 1 * 1 items
Catalog # MEMMB32191UF750TS
Supplier:  MEMMERT
Frequently Bought Together