Hybrid multi-parameter meter, handheld, SL1000 PPA
Product Details & Documents
The SL1000 is a portable colorimeter that provides 75% faster testing by using Chemkey® reagents instead of powder pillows or cuvette test methods. No reagent handling is necessary − all chemical and processes are entirely contained inside the Chemkey®. Besides that, the meter can be used in combination with digital IntelliCAL™ electrodes to measure different electrochemical parameters, such as pH, conductivity, ORP, dissolved oxygen, nitrate, fluoride in clean water applications.
- Ideal for disinfection control in the field or clean water treatment
- Automatically identifies the type of Chemkeys® or the type of electrode that is installed/connected
- Automation and internal temperature control for a consistent and repeatable process
- Meter can use a maximum of four Chemkeys® and a maximum of two electrodes for measurements
- Up to six parameters can be tested in parallel
Colorimetric tests: Copper, iron (dissolved), ortho-phosphate, hardness, alkalinity, nitrite, chlorine free, chlorine total and monochloramine.
Electrode measured parameters: pH, conductivity, ORP, dissolved oxygen, nitrate, fluoride (temperature is included with each electrode).
: The SL1000 PPA basic instrument kit is supplied in a carrying case with one SL1000 meter, two probe sample cups, one instrument sample cup, reachargeable battery, battery charger, handstrap, manual and USB cable.
The SL1000 PPA field kit includes the basic instrument kit plus one pH electrode, one conductivity electrode and one box each of free chlorine, total chlorine, monochloramine, nitrite and free ammonia Chemkeys®.
Specifications
- Description:Multi-parameter meter, SL1000 PPA (without Chemkey® reagents)
- Seq:001
- Connections:2× M12 connectors for IntelliCal™ probes, 4× Chemkey slots, 2,5 mm Jack for external power supply
- Cat. No.:705-0941
- Compliance:IP 64 with caps installed for the meter probe connector ports or with the probes connected to the instrument.
- Data logging - log function:Automatic in Press to Read measurement mode; manual in Continuous measurement mode
- Model:SL1000
- Outputs:Mini USB
- W×D×H:131×59×258 mm
- Data logging capacity:1000 measured values (result, date, time, site ID, user ID)
- Power supply:Lithium ion rechargeable battery or external charger (100 - 240 V, 50/60 Hz)
- Temperature compensation:Off, automatic and manual (for IntelliCal™ probes; parameter dependent)
- Weight:1,2 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement hand strap
Product Information
- Catalog #AvailabilityPrice
- System verification Chemkey®, verifies system functionality for testing all available parameters in less than 60 seconds (shelf life 12 months)System verification Chemkey®, verifies system functionality for testing all available parameters in less than 60 seconds (shelf life 12 months)705-0962Log in for availability1 * 1 items - £37.00
Product Information
- Catalog #AvailabilityPrice
- Car charger
Product Information
- Catalog #AvailabilityPrice
- Chlorine verification Chemkey®, verifies system functionality for testing chlorine in less than 60 secondsChlorine verification Chemkey®, verifies system functionality for testing chlorine in less than 60 seconds705-0961Log in for availability1 * 1 items - £26.80
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



