ContactUsLinkComponent
Resolution Life Science PC Software - includes Quick Read, Quick Scan,Fixed Wavelength, Wavelength Scan, Quantitative Analysis, Kine tics, Method Developer and Validation modules plus pre-stored life science methods. 1 * 1 items
Catalog # WAPR80-7100-32
Supplier:  Biochrom
Resolution Life Science PC Software - includes Quick Read, Quick Scan,Fixed Wavelength, Wavelength Scan, Quantitative Analysis, Kine tics, Method Developer and Validation modules plus pre-stored life science methods. 1 * 1 items
Catalog # WAPR80-7100-32
Supplier:  Biochrom
Frequently Bought Together