Resolution PC software - includes Quick Read, Quick Scan, Fixed Wavelength, Wavelength Scan, Quantitative Analysis, Kinetics, Method Developer and Validation modules. 1 * 1 items
Catalog # WAPR80-7100-31
Supplier: Biochrom
Resolution PC software - includes Quick Read, Quick Scan, Fixed Wavelength, Wavelength Scan, Quantitative Analysis, Kinetics, Method Developer and Validation modules. 1 * 1 items
Catalog # WAPR80-7100-31
Supplier: Biochrom