Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
1,8-Octanedithiol 99%

1,8-Octanedithiol 99%

Glass bottle, 1 g
Catalog # ACRO310620010
Irritant
1,8-Octanedithiol 99%
Catalog # ACRO310620010
CAS Number:  1191-62-4
1,8-Octanedithiol 99%
1,8-Octanedithiol 99%

Glass bottle, 1 g

CAS Number:  1191-62-4
Price
£61.90
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1,8-Dimercaptooctane
  • CAS Number:1191-62-4
  • Formula:C₈H₁₈S₂
  • MDL Number:MFCD00003574
  • Molecular Weight:178.36 g/mol
  • Boiling Pt:269…270 °C (760 mmHg)
  • Density:0,96 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:214-738-8
  • UN:3334
  • ADR:9,III
  •  :Density: 0.9814 g/cm³ (0 °C)

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5030 to 1.5050 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.