N-Methylaniline 99%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:100-61-8
- Formula:C₇H₉N
- MDL Number:MFCD00008283
- Molecular Weight:107.16 g/mol
- Merck Index:12,06098
- Boiling Pt:195 °C (760 mmHg)
- Melting Pt:–57 °C
- Density:0,987 g/cm³
- Flash Pt:86 °C
- Storage Temperature:Ambient
- EINECS:202-870-9
- UN:2294
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5685 to 1.5725 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.003 %
- Carbonyl compounds:≤0.01 % (as formaldehyde)
- Titrable acid:≤0.001 meq/g
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



