2,3-Dimethoxybenzaldehyde 97%
Glass bottle, 10 g
Supplier: Thermo Fisher Scientific
2,3-Dimethoxybenzaldehyde 97%
Catalog # ACRO115370100
Supplier: Thermo Fisher Scientific
CAS Number: 86-51-1
Jump to:
Product Details & Documents
- CAS Number:86-51-1
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003309
- Molecular Weight:166.18 g/mol
- Boiling Pt:137 °C (12 mmHg)
- Melting Pt:49…54 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:201-677-7
Documents
ACRO11537
SDS
Specifications
Specification Test Results
- Appearance:Off-white to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:48 to 53 °C
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



