2-Hydrazinylpyridine 98%
Glass bottle, 5 g
: 2(1H)-Pyridinone, hydrazone, 2-hydrazinylpyridine, 2-Hydrazinopyridine
- :4930-98-7
- :C₅H₇N₃
- :MFCD00006249
- :109.13 g/mol
- :185 °C (140 mmHg)
- :44,5…46 °C
- :110 °C
- :Refrigerator
- Appearance:White to orange-brown Crystals or crystalline mass
- Infrared spectrum:Conforms
- Melting point:43 to 47 °C
- GC:≥97.5 %
- Pack type:Glass bottle
- Pk:5 g



