ContactUsLinkComponent
Acid reagent cartridge for Sievers M-Series, 900, 5310 C, and 800 Series. Replacement recommended after 6 months. 1 * 1 items
Catalog # IONIAPF90310-02
Supplier:  IONICS
Acid reagent cartridge for Sievers M-Series, 900, 5310 C, and 800 Series. Replacement recommended after 6 months. 1 * 1 items
Catalog # IONIAPF90310-02
Supplier:  IONICS