Barium chloride in aqueous solution USP Test Solution (TS)
Plastic bottle, 500 mL
Supplier: VWR Chemicals
Barium chloride in aqueous solution USP Test Solution (TS)
Catalog # 85332.260
Supplier: VWR Chemicals
CAS Number: 10361-37-2
Jump to:
Product Details & Documents
Ready to use test/reagent solutions (TS and RS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
- CAS Number:10361-37-2
- Formula:BaCl₂
- MDL Number:MFCD00003445
- Molecular Weight:208.24 g/mol
- Merck Index:12,00998
- Melting Pt:~ 963 °C
- Storage Temperature:Ambient
- EINECS:0
- UN:3287
- ADR:6.1,III
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL



