Barium chloride in aqueous solution USP Test Solution (TS)
Plastic bottle, 100 mL
Supplier: VWR Chemicals
Barium chloride in aqueous solution USP Test Solution (TS)
Catalog # 85332.180
Supplier: VWR Chemicals
CAS Number: 10361-37-2
Jump to:
Product Details & Documents
Ready to use test/reagent solutions (TS and RS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
- CAS Number:10361-37-2
- Formula:BaCl₂
- MDL Number:MFCD00003445
- Molecular Weight:208.24 g/mol
- Merck Index:12,00998
- Melting Pt:~ 963 °C
- Storage Temperature:Ambient
- EINECS:0
- UN:3287
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



