ContactUsLinkComponent
Purified TFRC (ABM83785.1, 89 a.a. - 760 a.a.) human recombinant protein with His-Flag-StrepII tag at N-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00007037-H02
Supplier:  Abnova
Purified TFRC (ABM83785.1, 89 a.a. - 760 a.a.) human recombinant protein with His-Flag-StrepII tag at N-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00007037-H02
Supplier:  Abnova