Pipette serum 3ml 150mm polyethylene graduations 0,5-3ml non-sterile 1 * 3.000 items
Catalog # GREI741301_3000
Supplier: Greiner Bio-One
Pipette serum 3ml 150mm polyethylene graduations 0,5-3ml non-sterile 1 * 3.000 items
Catalog # GREI741301_3000
Supplier: Greiner Bio-One
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Transfer pipettes
Supplier: RATIOLAB
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Microplates, 96-well
Supplier: Greiner Bio-One
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection