ContactUsLinkComponent
safety storage cabinet, tall cabinet with folding doors, model line q-line, model class q-phoenix-90, model q90.195.120.fd, with cyl inder locking and locking state indicator dimensions: exterior: wxdxh (mm) 1200 x 615 x 1955 interior: wxdxh (mm) 105 1 * 1 items
  MSGA32149040
 :  asecos
safety storage cabinet, tall cabinet with folding doors, model line q-line, model class q-phoenix-90, model q90.195.120.fd, with cyl inder locking and locking state indicator dimensions: exterior: wxdxh (mm) 1200 x 615 x 1955 interior: wxdxh (mm) 105 1 * 1 items
  MSGA32149040
 :  asecos
Frequently Bought Together