ContactUsLinkComponent
erricans High density polyethylene 25 Ltr (brimful 28.2 ltrs). UN approved. UN mark 3H1/Y1.9/170/*/B/1382-040068 dimensions H468 x W260 x L296 + loose fitted tamper evident DIN 61 caps with epe foam gasket. 1 * 1 items
  216-1626
 :  Azpack
undefined
erricans High density polyethylene 25 Ltr (brimful 28.2 ltrs). UN approved. UN mark 3H1/Y1.9/170/*/B/1382-040068 dimensions H468 x W260 x L296 + loose fitted tamper evident DIN 61 caps with epe foam gasket. 1 * 1 items
  216-1626
 :  Azpack
Frequently Bought Together