TDBA 4-(3-TRIFLUOROMETHYL)-3H-DIAZIRIN-3 1 * 25 mg
Catalog # 8.51093.8025
Supplier: Novabiochem (Part of Merck)
TDBA 4-(3-TRIFLUOROMETHYL)-3H-DIAZIRIN-3 1 * 25 mg
Catalog # 8.51093.8025
Supplier: Novabiochem (Part of Merck)
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Test tubes
Supplier: Greiner Bio-One
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Cell culture storage bottles, polystyrene, Corning®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection