Potassium Hexacyanoferrate(II) 3-hydrate (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131505.1211
Supplier: PanReac AppliChem
Potassium Hexacyanoferrate(II) 3-hydrate (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131505.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW