Copper(II) Sulfate 5-hydrate (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131270.1211
Supplier: PanReac AppliChem
Copper(II) Sulfate 5-hydrate (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131270.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Universal Pipette Tips
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Eyewash solution, pH neutral
Supplier: Plum Safety
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Pasteur pipettes, disposable
Supplier: MARIENFELD
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Nitric Acid 69% for analysis, ACS, ISO 1 * 1.000 mL
Supplier: PanReac AppliChem
WIN-55,212-2 ≥98% (by HPLC)
Supplier: Enzo Life Sciences