Silver Nitrate 0.1 mol/l (0.1N) volumetric solution 1 * 2,5 L
APPC181464.1212
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Hydrochloric acid 37% Ph. Eur.
: VWR Chemicals