Citric Acid 1-hydrate for analysis, ACS, ISO 1 * 5 kg
Catalog # APPC131018.1214
Supplier: PanReac AppliChem
Citric Acid 1-hydrate for analysis, ACS, ISO 1 * 5 kg
Catalog # APPC131018.1214
Supplier: PanReac AppliChem
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate
Supplier: MACHEREY-NAGEL
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals