Drive belt (Hei-VAP Bench-top) 1 * 1 items
Catalog # HEID26-300-001-11
Supplier: Heidolph Instruments GmbH & Co.KG
Drive belt (Hei-VAP Bench-top) 1 * 1 items
Catalog # HEID26-300-001-11
Supplier: Heidolph Instruments GmbH & Co.KG
Frequently Bought Together
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Filter protection 5 ml 1 * 50 items
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for accu-jet® pro
Supplier: Brand
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals