ContactUsLinkComponent
EDTA disodium salt is a reversible metalloprotease inhibitor that is used for chelating metal ions. A 0.5M solution is also availabl e. 1 * 250 g
Catalog # GENORC-051
Supplier:  G-Biosciences
EDTA disodium salt is a reversible metalloprotease inhibitor that is used for chelating metal ions. A 0.5M solution is also availabl e. 1 * 250 g
Catalog # GENORC-051
Supplier:  G-Biosciences
Frequently Bought Together