ContactUsLinkComponent
G-Biosciences Immobilized Soybean Trypsin Inhibitor (STI) resin is designed for the efficient removal of trypsin, chymotrypsin and e lastase proteases from protein digests. The action of the Immobilized STI resin will stop enzymatic reactions. 1 * 2 mL
Catalog # GENO786-843
Supplier:  G-Biosciences
G-Biosciences Immobilized Soybean Trypsin Inhibitor (STI) resin is designed for the efficient removal of trypsin, chymotrypsin and e lastase proteases from protein digests. The action of the Immobilized STI resin will stop enzymatic reactions. 1 * 2 mL
Catalog # GENO786-843
Supplier:  G-Biosciences