ContactUsLinkComponent
DNAmark™ 50bp Plus Ladder consists of 13 fragments ranging from 50‐1000 base pairs (bp). This molecular weight marker is specially d esigned for identifying small DNA fragments and allowing for quantitative estimation of the DNA fragments in the gel 1 * 50 µG
Catalog # GENO786-853
Supplier:  G-Biosciences
DNAmark™ 50bp Plus Ladder consists of 13 fragments ranging from 50‐1000 base pairs (bp). This molecular weight marker is specially d esigned for identifying small DNA fragments and allowing for quantitative estimation of the DNA fragments in the gel 1 * 50 µG
Catalog # GENO786-853
Supplier:  G-Biosciences