ContactUsLinkComponent
TBST is commonly used as a wash solution for Western blot membranes and microtiter plate wells in ELISA assays. The Tris Buffered Sa line with Tween® 20 is an optimal formulation of pH stabilizers, salts and detergents designed to effectively remove 1 * 1 L
Catalog # GENOR042
Supplier:  G-Biosciences
TBST is commonly used as a wash solution for Western blot membranes and microtiter plate wells in ELISA assays. The Tris Buffered Sa line with Tween® 20 is an optimal formulation of pH stabilizers, salts and detergents designed to effectively remove 1 * 1 L
Catalog # GENOR042
Supplier:  G-Biosciences