G-Biosciences' Protein G Magnetic Beads are Fe3O4 beads coated with dextran to produce highly uniform, 3µm beads. Recombinant Prote in G is covalently coupled to the dextran coat to produce an enhanced alternative to agarose slurries for immunopreci 1 * 5 mL
Catalog # GENO786-905
Supplier: G-Biosciences
Frequently Bought Together