ContactUsLinkComponent
Carbonyl groups, including aldehydes, ketones and glyoxals, react with amines to form Schiff base intermediates that are in equilibr ium with their free forms. The labile Schiff’s base interaction can be stabilized by chemical reduction. 1 * 2.000 mg
Catalog # GENO786-062
Supplier:  G-Biosciences
Carbonyl groups, including aldehydes, ketones and glyoxals, react with amines to form Schiff base intermediates that are in equilibr ium with their free forms. The labile Schiff’s base interaction can be stabilized by chemical reduction. 1 * 2.000 mg
Catalog # GENO786-062
Supplier:  G-Biosciences